Learn More
Bio X Cell Antibody, Anti-human PD-1, IgG4, 5 MG
Supplier: Bio X Cell SIM0003R005MG
This non-therapeutic biosimilar antibody, derived from Nivolumab, reacts with human PD-1 (CD279). It is a humanized IgG4 monoclonal antibody engineered with a hinge mutation (S228P) to reduce non-specific binding and enhance stability for research applications.
- Isotype: Human IgG4; Immunogen: Human PD-1
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Purity: ≥95%; Endotoxin: ≤0.5EU/mg; Aggregation: <5%
- Purification: Protein A; Molecular weight: 150 kDa
- Applications: In vitro PD-1 neutralization, in vivo PD-1/PD-L signaling blockade, functional assays, Flow Cytometry, IHC, Western Blot
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.