Learn More
Bio X Cell InVivoSIM anti-human PD-1 Biosimilar, Human IgG4, 150 kDa, 1 MG
Supplier: Bio X Cell SIM0010R001MG
This non-therapeutic InVivoSIM anti-human PD-1 (Pembrolizumab Biosimilar) antibody is designed for research applications. It utilizes the same variable regions as the therapeutic antibody Pembrolizumab and specifically reacts with human PD-1 (programmed death-1), also known as CD279. PD-1 is a cell surface receptor involved in regulating T-cell activation, making this biosimilar valuable for studies on peripheral tolerance, autoimmune diseases, and cancer immunotherapies.
- Reacts with human PD-1
- Human IgG4 isotype
- 150 kDa molecular weight
- Purity of 95%
- Endotoxin level of 0.5 EU/mg
- Supports in vivo and in vitro PD-1/PD-L signaling blockade and functional assays
- Unconjugated for flexible experimental design
- Purified from cell culture supernatant in an animal-free facility
- Store at 4C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.