Learn More
Bio X Cell InVivoSIM Antibody, Anti-human PD-1, Pembrolizumab, 100 MG
Supplier: Bio X Cell SIM0010R100MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as Pembrolizumab, suitable for research. It reacts with human PD-1 (CD279), a cell surface receptor involved in negatively regulating TCR signals and preventing autoimmune diseases.
- Clone: Pembrolizumab
- Reactivity: Human
- Isotype: Human IgG4
- Immunogen: Human PD-1
- Applications: PD-1/PD-L signaling blocking, functional assays
- Formulation: PBS, pH 7.0 (preservative-free)
- Purity: ≥95% (Protein A purified)
- Sterility: 0.2 μm filtered
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.