Learn More
Bio X Cell Anti-PD-1 Antibody, Pembrolizumab, IgG4, 5 MG
Supplier: Bio X Cell SIM0010R005MG
This non-therapeutic biosimilar antibody, a Pembrolizumab biosimilar, reacts with human PD-1 (programmed death-1, CD279). It utilizes the same variable regions as the therapeutic antibody Pembrolizumab, making it ideal for research applications.
- Clone: Pembrolizumab
- Reactivities: Human
- Isotype: Human IgG4
- Conjugation: Unconjugated
- Immunogen: Human PD-1
- Reported applications: In vivo blocking of PD-1/PD-L signaling, in vitro blocking of PD-1/PD-L signaling, in vitro functional assay
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.