Learn More
Bio X Cell Ramucirumab Biosimilar, anti-human VEGFR-2, IgG1, 100 MG
Supplier: Bio X Cell SIM0012R100MG
This InVivoSIM anti-human VEGFR-2 (Ramucirumab Biosimilar) is a human IgG1 antibody developed for functional assays, immunohistochemistry, and flow cytometry, targeting human VEGFR2.
- Antibody host: human
- Species reactivity: human
- Antibody isotype: IgG1
- Immunogen: human VEGFR2
- Applications include: functional assays, immunohistochemistry, and flow cytometry
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.