Learn More
Bio X Cell Antibody, Ramucirumab VEGFR-2, 25 MG
Supplier: Bio X Cell SIM0012R025MG
This non-therapeutic biosimilar antibody, derived from Ramucirumab, is for research use. It reacts with human VEGFR-2 (CD309, KDR, Flk-1), which is crucial for vascular development. Ramucirumab binds to VEGFR-2, blocking VEGF ligands and inhibiting tumor angiogenesis.
- Isotype: human IgG1
- Immunogen: human VEGFR2
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 µm filtration
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Applications: functional assays, immunohistochemistry, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.