Learn More
Bio X Cell InVivoSIM anti-human CD20, Human IgG1, 25 MG
Supplier: Bio X Cell SIM0008R025MG
InVivoSIM anti-human CD20 (Rituximab Biosimilar) is designed for various in vivo and in vitro functional assays, including CD20+ cell depletion, flow cytometry, ELISA, and Western Blot.
- Antibody host: human
- Species reactivity: human
- Antibody isotype: IgG1
- Immunogen: human lymphoblastoid cell line SB
- Storage temperature: 4°C, do not freeze
- Shipping temperature: shipped on blue ice to maintain 4°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.