Learn More
Bio X Cell InVivoSIM Antibody, Anti-IL-17A, Secukinumab, IgG1, 25 MG
Supplier: Bio X Cell SIM0013R025MG
This InVivoSIM anti-human IL-17A antibody is a non-therapeutic biosimilar, utilizing Secukinumab's variable regions for research. It reacts with human IL-17A, a cytokine important in immunoregulation, inflammation, and autoimmune disease.
- Isotype: Human IgG1
- Reactivity: Human
- Immunogen: Human IL-17A
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg; Aggregation: <5%
- Purity: ≥95%; Sterility: 0.2 µm filtration
- Production: Purified from cell culture supernatant in animal-free facility
- Purification method: Protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Applications: In vitro IL-17A neutralization, functional assays, immunofluorescence, immunohistochemistry, flow cytometry, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.