Learn More
Bio X Cell Secukinumab Biosimilar, Human IL-17A, IgG1, 5 MG
Supplier: Bio X Cell SIM0013R005MG
This non-therapeutic biosimilar antibody shares the same variable regions as therapeutic Secukinumab, making it ideal for research. It reacts with human IL-17A, a cytokine involved in immunoregulation, inflammation, and autoimmune diseases.
- Clone: Secukinumab
- Reactivities: Human
- Isotype: Human IgG1
- Immunogen: Human IL-17A
- Applications: In vitro IL-17A neutralization, functional assays, immunofluorescence, IHC, flow cytometry, ELISA
- Formulation: PBS, pH 7.0, no stabilizers/preservatives
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.