Learn More
Bio X Cell Antibody, anti-human IL-17A, Secukinumab Biosimilar, 50 MG
Supplier: Bio X Cell SIM0013R050MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Secukinumab, making it suitable for research applications. It reacts with human IL-17A, a cytokine involved in immunoregulation and inflammation, playing a role in anti-microbial and chronic inflammation as well as autoimmune diseases. Secukinumab functions by neutralizing IL-17A.
- Isotype: human IgG1
- Recommended isotype control: RecombiMAb human IgG1 isotype control
- Immunogen: human IL-17A
- Reported applications: in vitro IL-17A neutralization, functional assays, immunofluorescence, immunohistochemistry, flow cytometry, ELISA
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.