Learn More
Bio X Cell anti-human IL-6R, Tocilizumab, Unconjugated, 100 MG
Supplier: Bio X Cell SIM0014R100MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Tocilizumab, making it suitable for research applications. It interacts with human IL-6 receptor (CD126), an 80 kDa type I cytokine receptor belonging to the immunoglobulin superfamily. It binds to both soluble and membrane-bound IL-6R, blocking the interaction of IL-6 with its receptor.
- Isotype: Human IgG1
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg), determined by LAL assay
- Sterility: 0.2 μm filtration
- Purification: Protein A
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Applications: Functional assays, ELISA
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.