Learn More
Bio X Cell InVivoSIM anti-human IL-6R, Clone Tocilizumab, 25 MG
Supplier: Bio X Cell SIM0014R025MG
This non-therapeutic biosimilar antibody, an InVivoSIM product, uses the same variable regions as the therapeutic antibody Tocilizumab, making it suitable for research. It reacts with human IL-6 receptor (CD126), influencing immune and inflammatory responses, neuronal development, and mediating acute phase reactions by blocking IL-6 binding to both soluble and membrane-bound IL-6R.
- Clone: Tocilizumab
- Isotype: Human IgG1
- Reported applications: Functional assays, ELISA
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.