Learn More
Aviva Systems Biology Corp SIM1 Antibody - N-terminal region (ARP38549_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38549_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIM1. Alternate names: bHLHe14ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56698