Learn More
Aviva Systems Biology Corp SIM2 Antibody - N-terminal region (ARP38551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIM2. Alternate names: MGC119447, SIM, bHLHe15, HMC13F06, HMC29C01ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56699