Learn More
Cayman Chemical Simazine-d10, Purity: ≥99% deuterated forms (d1-d10), CAS: 220621-39-6, 10 mg
SDP
Catalog No. 502910907
Encompass_Preferred
An internal standard for the quantification of simazine by GC- or LC-MS
Catalog No. 50-291-0907 Supplier Cayman Chemical Supplier No. 3520910 MG
An internal standard for the quantification of simazine by GC- or LC-MS