Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Simazine | 122-34-9 | 99.4% | 1 ML
SDP

Catalog No. 5000409847
Encompass_Preferred

Simazine is a triazine herbicide widely used in agriculture and plant production. It induces apoptosis of immune cells and inhibits B and T cell proliferation in mice. Simazine can also disrupt spindle morphology, affect mitochondrial function, increase ROS, and induce DNA damage and early apoptosis in mouse oocytes. In vivo, it leads to immunosuppression and immunotoxicity in mice.

  • Target: Triazine herbicide.
  • Biological activity: Induces immune cell apoptosis; inhibits B and T cell proliferation.
  • In vitro effects: Inhibits oocyte maturation; disrupts spindle; affects mitochondrial function; increases ROS; induces DNA damage and apoptosis.
  • In vivo effects: Causes immunosuppression and immunotoxicity; reduces spleen T and B cell proliferation.
  • Storage: 4°C, protected from light. In solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-004-09847 Supplier Medchemexpress LLC Supplier No. HYB204610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simazine is a triazine herbicide widely used in agriculture and plant production. It induces apoptosis of immune cells and inhibits B and T cell proliferation in mice. Simazine can also disrupt spindle morphology, affect mitochondrial function, increase ROS, and induce DNA damage and early apoptosis in mouse oocytes. In vivo, it leads to immunosuppression and immunotoxicity in mice.

  • Target: Triazine herbicide.
  • Biological activity: Induces immune cell apoptosis; inhibits B and T cell proliferation.
  • In vitro effects: Inhibits oocyte maturation; disrupts spindle; affects mitochondrial function; increases ROS; induces DNA damage and apoptosis.
  • In vivo effects: Causes immunosuppression and immunotoxicity; reduces spleen T and B cell proliferation.
  • Storage: 4°C, protected from light. In solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.