Learn More
Medchemexpress LLC Simazine | 122-34-9 | 99.4% | 100 MG
SDP
Catalog No. 5000411626
Encompass_Preferred
Simazine is a triazine herbicide widely used in agriculture, potted plant, and tree production. It can induce apoptosis of immune cells in the spleen of mice and inhibit the proliferation of B cells and T cells in mice.
  • Triazine herbicide
  • Can inhibit maturation, disrupt spindle morphology, affect mitochondrial function, increase ROS levels, and induce DNA damage and early apoptosis of mouse oocytes
  • Can lead to immunosuppression and immunotoxicity in mice
  • Reduced proliferation of both spleen T and B cells in mice
  • Decreased the number of spleen CD4+ T lymphocytes, while CD8+ T cells remained unchanged in mice
  • Reduced immune function, higher intracellular [Ca2+], and oxidative stress in mice
  • Induced spleen immune cell apoptosis by reducing Bcl-2, while increasing Fas and Caspase-3 levels in mice
  • Appearance: Solid, white to off-white
Catalog No. 50-004-11626 Supplier Medchemexpress LLC Supplier No. HYB2046100MG
Simazine is a triazine herbicide widely used in agriculture, potted plant, and tree production. It can induce apoptosis of immune cells in the spleen of mice and inhibit the proliferation of B cells and T cells in mice.
  • Triazine herbicide
  • Can inhibit maturation, disrupt spindle morphology, affect mitochondrial function, increase ROS levels, and induce DNA damage and early apoptosis of mouse oocytes
  • Can lead to immunosuppression and immunotoxicity in mice
  • Reduced proliferation of both spleen T and B cells in mice
  • Decreased the number of spleen CD4+ T lymphocytes, while CD8+ T cells remained unchanged in mice
  • Reduced immune function, higher intracellular [Ca2+], and oxidative stress in mice
  • Induced spleen immune cell apoptosis by reducing Bcl-2, while increasing Fas and Caspase-3 levels in mice
  • Appearance: Solid, white to off-white