Learn More
Medchemexpress LLC Simazine | 122-34-9 | 99.4% | 50 MG
SDP
Catalog No. 5000409848
Encompass_Preferred
Simazine is a triazine herbicide widely used in agriculture, potted plant, and tree production. It can also induce apoptosis of immune cells in the spleen of mice and inhibit the proliferation of B cells and T cells in mice.
  • Inhibits maturation and disrupts spindle morphology in mouse oocytes
  • Affects mitochondrial function and increases ROS levels
  • Induces DNA damage and early apoptosis of mouse oocytes
  • Leads to immunosuppression and immunotoxicity in mice
  • Reduces proliferation of spleen T and B cells
  • Decreases numbers of spleen CD4+ T lymphocytes
  • Reduces immune function and causes higher intracellular [Ca2+] and oxidative stress
Catalog No. 50-004-09848 Supplier Medchemexpress LLC Supplier No. HYB204650MG
Simazine is a triazine herbicide widely used in agriculture, potted plant, and tree production. It can also induce apoptosis of immune cells in the spleen of mice and inhibit the proliferation of B cells and T cells in mice.
  • Inhibits maturation and disrupts spindle morphology in mouse oocytes
  • Affects mitochondrial function and increases ROS levels
  • Induces DNA damage and early apoptosis of mouse oocytes
  • Leads to immunosuppression and immunotoxicity in mice
  • Reduces proliferation of spleen T and B cells
  • Decreases numbers of spleen CD4+ T lymphocytes
  • Reduces immune function and causes higher intracellular [Ca2+] and oxidative stress