Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simazine Solution, 100μg/mL in acetone, Ultra Scientific

Catalog No. USPP530A1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPP530A1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPP530A1 Supplier Agilent Technologies Supplier No. PP530A1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-34-9 , 67-64-1
Molecular Formula C7H12ClN5
Molecular Weight (g/mol) 201.658
InChI Key ODCWYMIRDDJXKW-UHFFFAOYSA-N
PubChem CID 5216
ChEBI CHEBI:27496
IUPAC Name 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(=NC(=N1)Cl)NCC

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Acetone,Simazine
Element Acetone, Simazine
Solvent or Matrix Acetone
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Simazine Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.