Learn More
3B Scientific™ SimBP Simulator for Blood Pressure Training
GREENER_CHOICESDPSCI_ED
Catalog No. S58112 Shop All 3B Scientific Products
Change view
Click to view available options
Blood Pressure Simulator
Catalog No.
S58112 Blood Pressure Simulator
1 options
This item is not returnable. View return policy
Catalog No. S58112 Supplier 3B Scientific™ Supplier No. 1022869
Add to Cart
Add to Cart
This item is not returnable. View return policy
Train the fundamental clinical skill of manual blood pressure measurement with the new SimBP™ by Cardionics. SimBP™ simulates a manual sphygmomanometer, a reliable instrument for measuring blood pressure to provide the most accurate readings of upper and lower pressure. Configurable features add an extra layer of realism to standardized patient scenarios as well as manikin-based simulation.
  • Students will benefit from the training possibilities of SimBP™ to perform standard blood pressure procedures with this sphygmomanometer simulator on either a standardized patient or a manikin. SimBP™ has a soft, flexible inflation cuff and an authentic bulb and gauge for even more life-like training scenarios.
  • SimBP™ simulates various blood pressure conditions, which can be easily controlled via the software.
  • Systolic pressure, diastolic pressure as well as heart rate can be manipulated.
  • By utilizing the SimScope™ stethoscope (included) students can listen to 5 Korotkoff phases and learn how to identify the Auscultatory gap.
  • The simulator can also be used by instructors to test the students' procedural skills for measuring systolic and diastolic values.
  • Blood pressure simulator with a high-quality gauge and a bulb to simulate blood pressure measurement on a standardized patient or a manikin for added realism
  • Soft and easy to put on cuff, with a flexible inflation bladder
  • Individual manipulation of systolic and diastolic pressures, as well as the heart rat
  • Measurements range between 20 and 300 mmHg
  • Reduced cuff pressure inflation for a more comfortable feel of the standardized patient
  • Adjustable volume
Specifications
Product Type Blood Pressure Simulator
For Use With (Application) To Train and Increase Compentecy and Confidence in Taking Blood Pressure at The Entry Level.