Learn More
Med Vet International 12mL Sekurus Self-Tenting injector
Catalog No. 502827788
Encompass_Preferred
Self-tenting Needle Guard ensures a greater level of user protection and is highly effective on a wide range of animal skin types. Needle change is faster and safer. Needle damage is significantly reduced, increasing the life of the needle. Safer operation in confined spaces such as chutes and cattle crushes. Dose is correctly administered in the subcutaneous zone. Less invasive to the animal.
Catalog No. 50-282-7788 Supplier Med Vet International Supplier No. PHVSEKURUS12ML
Self-tenting Needle Guard ensures a greater level of user protection and is highly effective on a wide range of animal skin types. Needle change is faster and safer. Needle damage is significantly reduced, increasing the life of the needle. Safer operation in confined spaces such as chutes and cattle crushes. Dose is correctly administered in the subcutaneous zone. Less invasive to the animal.