Learn More
FUJIFILM Biosciences Simeconazole Standard
Catalog No. 5000450422
Encompass_Preferred
High-purity standard 99% of Simeconazole a triazole fungicide intended for pesticide residue analysis via GC and qNMR Supplied as a crystalline powder and suitable for analytical applications in food safety and environmental testing
Catalog No. 50-004-50422 Supplier FUJIFILM Biosciences Supplier No. 19612781
High-purity standard 99% of Simeconazole a triazole fungicide intended for pesticide residue analysis via GC and qNMR Supplied as a crystalline powder and suitable for analytical applications in food safety and environmental testing