Learn More
TARGETMOL CHEMICALS INC SIMEPREVIR 50MG
SDP
Catalog No. 502576734
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Simeprevir (TMC435) is a potent HCV NS3/4A protease inhibitor and inhibits HCV replication with EC50 of 8 nM. purity: 99%
Catalog No. 50-257-6734 Supplier TARGETMOL CHEMICALS INC Supplier No. T468650MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Simeprevir (TMC435) is a potent HCV NS3/4A protease inhibitor and inhibits HCV replication with EC50 of 8 nM. purity: 99%