Learn More
Medchemexpress LLC Simeprevir | 923604-59-5 | 98.79% | 749.94 | 100 MG
SDP
Catalog No. 5000364915
Encompass_Preferred
Simeprevir (TMC435) is an oral, potent, and highly specific hepatitis C virus (HCV) NS3/4A protease inhibitor. It inhibits HCV replication and also potently suppresses SARS-CoV-2 replication, synergizing with Remdesivir. It inhibits the main protease (Mpro) and the RNA-dependent RNA polymerase (RdRp) of SARS-CoV-2, and modulates host immune responses.
  • Oral, potent, and highly specific inhibitor
  • Inhibits HCV NS3/4A protease
  • Suppresses SARS-CoV-2 replication
  • Synergizes with Remdesivir
  • Modulates host immune responses
Catalog No. 50-003-64915 Supplier Medchemexpress LLC Supplier No. 100MGHY10241
Simeprevir (TMC435) is an oral, potent, and highly specific hepatitis C virus (HCV) NS3/4A protease inhibitor. It inhibits HCV replication and also potently suppresses SARS-CoV-2 replication, synergizing with Remdesivir. It inhibits the main protease (Mpro) and the RNA-dependent RNA polymerase (RdRp) of SARS-CoV-2, and modulates host immune responses.
  • Oral, potent, and highly specific inhibitor
  • Inhibits HCV NS3/4A protease
  • Suppresses SARS-CoV-2 replication
  • Synergizes with Remdesivir
  • Modulates host immune responses