Learn More
Cayman Chemical Simeprevir (sodium salt), Purity: ≥98%, CAS: 1241946-89-3, 10 mg
SDP
Catalog No. 502986228
Encompass_Preferred
A potent NS3/4A protease inhibitor (Ki = 0.36 nM); inhibits HCV viral replication (EC50 = 7.8 nM); effective against the HCV genotypes 1a and 1b in biochemical assays; works synergistically with IFN-α or NM-107 in replicon cells; works additively with ribavirin in replicon cells
Catalog No. 50-298-6228 Supplier Cayman Chemical Supplier No. 2214410 MG
A potent NS3/4A protease inhibitor (Ki = 0.36 nM); inhibits HCV viral replication (EC50 = 7.8 nM); effective against the HCV genotypes 1a and 1b in biochemical assays; works synergistically with IFN-α or NM-107 in replicon cells; works additively with ribavirin in replicon cells