Learn More
Medchemexpress LLC Simethicone | 8050-81-5 | 96.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10951950G
Encompass_Preferred
Simethicone is an orally active defoamer that reduces the surface tension of air bubbles in the gastrointestinal tract, leading to their expulsion by vomiting, exhalation, or absorption into the bloodstream. It has potential applications in treating flatulence and colic.
  • Reduces surface tension of air bubbles
  • Facilitates expulsion of gas
  • Potential application in flatulence
  • Potential application in colic
  • 96.00% assay
  • Liquid appearance
  • Colorless to light yellow
Catalog No. 50-003-44700