Learn More
AdipoGen Simetone
SDP
Catalog No. 502560070
Encompass_Preferred
Chemical. CAS 673-04-1. Formula C8H15N5O. MW 197.3. Synthetic. Triazine herbicide. Mode of action through inhibition of photosynthesis at photosystem II. Compound can be used as analytical reference material.
Catalog No. 50-256-0070 Supplier AdipoGen Supplier No. CDXS0057M025
Chemical. CAS 673-04-1. Formula C8H15N5O. MW 197.3. Synthetic. Triazine herbicide. Mode of action through inhibition of photosynthesis at photosystem II. Compound can be used as analytical reference material.