Learn More
Biomatik Corporation Simian Angiotensin I (AngI) ELISA Kit
Supplier:  Biomatik Corporation EKU11930
Encompass_Preferred
Simian Angiotensin I (AngI) ELISA Kit for detecting AngI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.84pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A2K5UDY1. Target Synonyms: Ang-I; Angiotensin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8063