Learn More
Biomatik Corporation Simian Atrial Natriuretic Peptide (ANP) ELISA Kit
Supplier:  Biomatik Corporation EKU11566
Encompass_Preferred
Simian Atrial Natriuretic Peptide (ANP) ELISA Kit for detecting ANP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.35pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A2K5VBM2. Gene ID: 102116318. Target Synonyms: ANH; ANF; CDD; Atrial Natriuretic Factor; Atrial Natriuretic Hormone; Atriopeptin; Cardionatrine; Cardiodilatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7507