Learn More
Biomatik Corporation Simian Beta-2-Microglobulin (b2M) ELISA Kit
Supplier:  Biomatik Corporation EKU08993
Encompass_Preferred
Simian Beta-2-Microglobulin (b2M) ELISA Kit for detecting b2M in serum, plasma, urine, other biological fluids. Species Reactivity: Simian. Detection Range: 15.6-1,000pg/ml. Sensitivity: 7.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6V7J5. Gene ID: 712428. Target Synonyms: BMG; B2MG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5426