Learn More
Biomatik Corporation Simian Carbonic Anhydrase II (CA2) ELISA Kit
Supplier:  Biomatik Corporation EKU02919
Encompass_Preferred
Simian Carbonic Anhydrase II (CA2) ELISA Kit for detecting CA2 in serum, plasma, other biological fluids. Species Reactivity: Simian. Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: H9FZI2. Target Synonyms: CA-II; CAII; Car2; CAC; Carbonate dehydratase II; Carbonic anhydrase C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9077