Learn More
Biomatik Corporation Simian Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKU04250
Encompass_Preferred
Simian Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Simian. Detection Range: 2.47-200ng/ml. Sensitivity: 0.95ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9BEH3; Q6EV79. Gene ID: 102138376; 101864888. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8972