Learn More
Biomatik Corporation Simian Interferon Gamma (IFNg) CLIA Kit
Supplier:  Biomatik Corporation EKU09749
Encompass_Preferred
Simian Interferon Gamma (IFNg) CLIA Kit for detecting IFNg in serum, plasma, other biological fluids. Species Reactivity: Simian. Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.57pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P63310. Gene ID: 574282. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7080