Learn More
Biomatik Corporation Simian Squamous Cell Carcinoma Antigen 1/2 (SCCA1/SCCA2) ELISA Kit
Supplier:  Biomatik Corporation EKU11973
Encompass_Preferred
Simian Squamous Cell Carcinoma Antigen 1/2 (SCCA1/SCCA2) ELISA Kit for detecting SCCA1/SCCA2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: SERPINB3; Serpin B3; SCCA1/SCCA2 fusion protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8057