Learn More
TARGETMOL CHEMICALS INC Simmitecan hydrochloride 1MG
SDP
Catalog No. 502878158
Encompass_Preferred
Also available in 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Simmitecan hydrochloride is a camptothecin derivative, a topoisomerase I inhibitor with anticancer activity, which can be used to study is solid tumors. Purity 98.36%
Catalog No. 50-287-8158 Supplier TARGETMOL CHEMICALS INC Supplier No. T711371MG
Also available in 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Simmitecan hydrochloride is a camptothecin derivative, a topoisomerase I inhibitor with anticancer activity, which can be used to study is solid tumors. Purity 98.36%