Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quanterix Inc SIMOA DISCS 16 PER STACK
SDP

Catalog No. NC1442827
Encompass

  • Product type: Simoa Discs
  • Quantity per stack: 16 discs
  • Compatible instruments: Simoa HD-1 Analyzer, Simoa HD-X Analyzer, SR-X System
  • Arrays per disc: 24 arrays
  • Microwells per array: 216,000 microwells
  • Technology: Single Molecule Array (Simoa)
  • Application: Ultra-sensitive immunoassay, Biomarker detection, In vitro diagnostics
  • Function: Enhances assay sensitivity and reproducibility
  • Typical use per kit: Enough discs for 4 plates
  • Supplier: Quanterix Inc
  • Disc manufacturing partner: Sony DADC

Catalog No. NC1442827 Supplier Quanterix Inc Supplier No. 100001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Product type: Simoa Discs
  • Quantity per stack: 16 discs
  • Compatible instruments: Simoa HD-1 Analyzer, Simoa HD-X Analyzer, SR-X System
  • Arrays per disc: 24 arrays
  • Microwells per array: 216,000 microwells
  • Technology: Single Molecule Array (Simoa)
  • Application: Ultra-sensitive immunoassay, Biomarker detection, In vitro diagnostics
  • Function: Enhances assay sensitivity and reproducibility
  • Typical use per kit: Enough discs for 4 plates
  • Supplier: Quanterix Inc
  • Disc manufacturing partner: Sony DADC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.