Learn More
Med Vet International Simparica Chewables For Dogs 44.1 To 88 Pounds, Green Label (6 Dose X 10)
Catalog No. 502828746
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Simparica (sarolaner) kills adult fleas, and is indicated for the treatment and prevention of flea infestations (Ctenocephalides felis), and the treatment and control of tick infestations [Amblyomma americanum (Lone Star tick), Amblyomma maculatum (Gulf Coast tick), Dermacentor variabilis (American dog tick), Ixodes scapularis (black-legged tick), and Rhipicephalus sanguineus (brown dog tick)] for one month in dogs 6 months of age or older and weighing 2.8 pounds or greater. See package insert for complete product details.
Catalog No. 50-282-8746 Supplier Med Vet International Supplier No. RXSIMPARICACH6GR
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Simparica (sarolaner) kills adult fleas, and is indicated for the treatment and prevention of flea infestations (Ctenocephalides felis), and the treatment and control of tick infestations [Amblyomma americanum (Lone Star tick), Amblyomma maculatum (Gulf Coast tick), Dermacentor variabilis (American dog tick), Ixodes scapularis (black-legged tick), and Rhipicephalus sanguineus (brown dog tick)] for one month in dogs 6 months of age or older and weighing 2.8 pounds or greater. See package insert for complete product details.