Learn More
Med Vet International Simparica Trio Chewable Tablets For Dogs 5.6 To 11 Pounds, Purple Label (6 Dose X 10)
Supplier:  Med Vet International RXSIMPARICATRIO6PU
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Simparica TRIO (sarolaner, moxidectin, and pyrantel) Chewable Tablets prevent heartworm disease caused by Dirofilaria immitis, kills adult fleas (Ctenocephalides felis) and is indicated for the treatment and prevention of flea infestations, the treatment and control of tick infestations with Amblyomma americanum (lone star tick), Amblyomma maculatum (Gulf Coast tick), Dermacentor variabilis (American dog tick), Ixodes scapularis (black-legged tick), and Rhipicephalus sanguineus (brown dog tick), and the treatment and control of roundworm (immature adult and adult Toxocara canis and adult Toxascaris leonina) and adult hookworm (Ancylostoma caninum and Uncinaria stenocephala) infections for one month in dogs and puppies 8 weeks of age and older, and weighing 2.8 pounds or greater. Simparica TRIO is indicated for the prevention of Borrelia burgdorferi infections
Catalog No. 50-282-8652