Learn More
Research Products International Corp Simplate Thin Wall PCR Plate, 96 Well, 250l Capacity, 50 per Case
SDP
Catalog No. 50212239
Encompass
Precision molded PCR plates ensure optimum uniformity and heat transfer. Thin, flexible design conforms to heating block wells and can be easily cut into smaller formats. Plates feature alphanumeric indexing, notched orientation corner and smooth top surface.
Plates accept various sealing materials such as films, micro-mats and caps. Natural color, autoclavable.
Catalog No. 50-212-239 Supplier Research Products International Corp Supplier No. 141314
Precision molded PCR plates ensure optimum uniformity and heat transfer. Thin, flexible design conforms to heating block wells and can be easily cut into smaller formats. Plates feature alphanumeric indexing, notched orientation corner and smooth top surface.
Plates accept various sealing materials such as films, micro-mats and caps. Natural color, autoclavable.