Learn More
MSC Simple Green Anti-Spatter, Oil and Silicone-Free, Non-Flammable, 1 gal.
Catalog No. 502868449
Encompass
Simple Green Anti-Spatter is a water-based, non-flammable solution designed to protect welding equipment and work surfaces from spatter. It is oil and silicone-free and contains no chlorinated solvents, making it suitable for use prior to coating, plating, or painting.
  • Protects welding equipment and work surfaces from spatter
  • Oil and silicone-free formula
  • No chlorinated solvents
  • Rinses easily, suitable before coating, plating, or painting
  • Water-based and non-flammable
  • Non-caustic, prevents harmful fumes when heated
  • Protects aluminum, steel, and other metal surfaces
  • Rinses residue-free
  • Ready-to-use, no mix formula
Catalog No. 50-286-8449 Supplier MSC Supplier No. 43384361
Simple Green Anti-Spatter is a water-based, non-flammable solution designed to protect welding equipment and work surfaces from spatter. It is oil and silicone-free and contains no chlorinated solvents, making it suitable for use prior to coating, plating, or painting.
  • Protects welding equipment and work surfaces from spatter
  • Oil and silicone-free formula
  • No chlorinated solvents
  • Rinses easily, suitable before coating, plating, or painting
  • Water-based and non-flammable
  • Non-caustic, prevents harmful fumes when heated
  • Protects aluminum, steel, and other metal surfaces
  • Rinses residue-free
  • Ready-to-use, no mix formula