Learn More
MSC Simple Green Anti Spatter, Liquid, Water-Based, Unscented, 55 gal
Catalog No. 502868543
Encompass
This anti-spatter solution comes in a 55-gallon drum and is a liquid, water-based, and unscented product. It is specifically formulated for the welding industry as a high-performance spatter release solution.
  • High-performance spatter release solution specially formulated for the welding industry
  • Safe for aluminum, steel and other metal surfaces
  • Ready-to-use formula goes on easily and quickly
  • Biodegradable with no added color or scent
  • Keeps nozzles, diffusers, tips and shrouds clear of build up
  • Helps to maintain optimum equipment performance and lifespan
Catalog No. 50-286-8543 Supplier MSC Supplier No. 43422906
This anti-spatter solution comes in a 55-gallon drum and is a liquid, water-based, and unscented product. It is specifically formulated for the welding industry as a high-performance spatter release solution.
  • High-performance spatter release solution specially formulated for the welding industry
  • Safe for aluminum, steel and other metal surfaces
  • Ready-to-use formula goes on easily and quickly
  • Biodegradable with no added color or scent
  • Keeps nozzles, diffusers, tips and shrouds clear of build up
  • Helps to maintain optimum equipment performance and lifespan