Learn More
MSC Simple Green Anti Spatter, Water Based, Unscented, 5 gal
Catalog No. 502867895
Encompass
Simple Green Anti Spatter is a liquid, water-based, and unscented solution specifically formulated for the welding industry. It is designed to be safe for aluminum, steel, and other metal surfaces, helping to maintain optimum equipment performance and lifespan.
  • High performance spatter release solution
  • Safe for aluminum, steel and other metal surfaces
  • Ready-to-use formula goes on easily and quickly
  • Biodegradable with no added color or scent
  • Keeps nozzles, diffusers, tips and shrouds clear of build up
  • Helps to maintain optimum equipment performance and lifespan
Catalog No. 50-286-7895 Supplier MSC Supplier No. 43384387
Simple Green Anti Spatter is a liquid, water-based, and unscented solution specifically formulated for the welding industry. It is designed to be safe for aluminum, steel, and other metal surfaces, helping to maintain optimum equipment performance and lifespan.
  • High performance spatter release solution
  • Safe for aluminum, steel and other metal surfaces
  • Ready-to-use formula goes on easily and quickly
  • Biodegradable with no added color or scent
  • Keeps nozzles, diffusers, tips and shrouds clear of build up
  • Helps to maintain optimum equipment performance and lifespan