Learn More
Enterprise Technology Solutions Simple Green Bathroom Cleaner Concentrate, 1 Gallon, Pink
SDP
Catalog No. 5000789968
Encompass_Preferred
Bathroom cleaner removes mineral-based soils, soap scum and hard water stains typically found on hard bathroom surfaces. The cleaner is noncaustic, nonflammable and formulated without VOCs that can adversely impact indoor air quality. Concentrated formula dilutes at 1:20 for great value. It meets the Green Seal GS-37 Standard, including reduced health, safety and environmental impact of formulations as well as product efficacy requirements. It also complies with LEED green building requirements and environmentally preferable product purchasing programs in place throughout federal, state and municipal governments.
Catalog No. 50-007-89968 Supplier Enterprise Technology Solutions Supplier No. SMP11101
Bathroom cleaner removes mineral-based soils, soap scum and hard water stains typically found on hard bathroom surfaces. The cleaner is noncaustic, nonflammable and formulated without VOCs that can adversely impact indoor air quality. Concentrated formula dilutes at 1:20 for great value. It meets the Green Seal GS-37 Standard, including reduced health, safety and environmental impact of formulations as well as product efficacy requirements. It also complies with LEED green building requirements and environmentally preferable product purchasing programs in place throughout federal, state and municipal governments.