Learn More
Enterprise Technology Solutions Simple Green Cleaner and Degreaser, 55 Gal, 23-1/5"Wx23-1/5"Lx35"H, Green
SDP
Catalog No. 5000790000
Encompass_Preferred
Industrial cleaner and degreaser is a powerful, industrial-strength cleaner and degreaser formulated for critical cleaning applications that demand high performance from a fragrance and color-free solution. This highly trusted cleaner utilizes a nonflammable formula to provide a great solution for a variety of industries where sanitation and safety are of equal concern. Clean-rinsing, concentrated and cost-effective formula is great for food processing and handling, industrial, manufacturing, electronics and parts washing industries. The formula is NSF labeled A1, A2, A4, A8, B1 and C1 for use in commercial kitchens, food-processing plants and other areas where food is prepared or handled. It is able to validate below 10 parts per million in rinsability requirements for cleaning in cosmetics, medical and pharmaceutical environments where Good Manufacturing Practices (GMP) are followed.
Catalog No. 50-007-90000 Supplier Enterprise Technology Solutions Supplier No. SMP13008
Industrial cleaner and degreaser is a powerful, industrial-strength cleaner and degreaser formulated for critical cleaning applications that demand high performance from a fragrance and color-free solution. This highly trusted cleaner utilizes a nonflammable formula to provide a great solution for a variety of industries where sanitation and safety are of equal concern. Clean-rinsing, concentrated and cost-effective formula is great for food processing and handling, industrial, manufacturing, electronics and parts washing industries. The formula is NSF labeled A1, A2, A4, A8, B1 and C1 for use in commercial kitchens, food-processing plants and other areas where food is prepared or handled. It is able to validate below 10 parts per million in rinsability requirements for cleaning in cosmetics, medical and pharmaceutical environments where Good Manufacturing Practices (GMP) are followed.