Learn More
MSC Simple Green Industrial Cleaner & Degreaser, Sassafras Scent, Concentrated, 1 Gallon
Catalog No. 502836853
Encompass
This industrial strength cleaner and degreaser provides powerful cleaning for a variety of surfaces and equipment. Recognized by the U.S. EPA's Safer Choice Program, its concentrated, water-based formula effectively removes dirt, grease, oil, and stains without harsh chemicals, making it a versatile and safer cleaning solution.
  • Industrial strength cleaning and degreasing
  • EPA Safer Choice certified
  • Removes dirt, grease, oil, and stains
  • Suitable for manual or professional cleaning
  • Rinses easily without residue
  • Non-flammable, reduces workplace hazards
  • Water-based, no butyl, solvents, or phosphates
  • Mild alkaline pH for broad application
  • Cost-effective concentrate with sassafras scent
  • Exempt from special handling requirements
Catalog No. 50-283-6853 Supplier MSC Supplier No. 63128524
This industrial strength cleaner and degreaser provides powerful cleaning for a variety of surfaces and equipment. Recognized by the U.S. EPA's Safer Choice Program, its concentrated, water-based formula effectively removes dirt, grease, oil, and stains without harsh chemicals, making it a versatile and safer cleaning solution.
  • Industrial strength cleaning and degreasing
  • EPA Safer Choice certified
  • Removes dirt, grease, oil, and stains
  • Suitable for manual or professional cleaning
  • Rinses easily without residue
  • Non-flammable, reduces workplace hazards
  • Water-based, no butyl, solvents, or phosphates
  • Mild alkaline pH for broad application
  • Cost-effective concentrate with sassafras scent
  • Exempt from special handling requirements