Learn More
Essendant d Pro 5 Disinfectant, 1 gal Bottle
Catalog No. NC9553286
Encompass_Preferred
Simple Green d Pro 5 One-Step Disinfectant is an EPA registered, hospital grade disinfectant known for its high performance. The highly versatile concentrate is a disinfectant, cleaner, sanitizer, fungicide, mildewstat, virucide and deodorizer. It kills HIV-1, Hepatitis B and C, SARS, Staph, MRSA, Pox Virus, Salmonella, E. Coli, Norovirus, Influenza, Athlete's Foot Fungus and other Dangerous and communicable health threats. It has no added color or scent making it ideal for food production, processing, and service facilities. Its strong cleaning ability and disinfecting power are perfect for high-traffic common areas such as restrooms, waiting rooms, lobbies and locker rooms.
This item is not returnable. View return policy
Catalog No. NC9553286 Supplier Essendant Supplier No. SMP30501CT
Simple Green d Pro 5 One-Step Disinfectant is an EPA registered, hospital grade disinfectant known for its high performance. The highly versatile concentrate is a disinfectant, cleaner, sanitizer, fungicide, mildewstat, virucide and deodorizer. It kills HIV-1, Hepatitis B and C, SARS, Staph, MRSA, Pox Virus, Salmonella, E. Coli, Norovirus, Influenza, Athlete's Foot Fungus and other Dangerous and communicable health threats. It has no added color or scent making it ideal for food production, processing, and service facilities. Its strong cleaning ability and disinfecting power are perfect for high-traffic common areas such as restrooms, waiting rooms, lobbies and locker rooms.