Learn More
MSC Simple Green Foaming Coil Cleaner, NSF Registered, 20 oz
Catalog No. 5000502405
Encompass
This professional-strength foaming coil cleaner penetrates quickly to remove dirt, grease, and grime that can clog coils and impede heat transfer. It is NSF registered for use in and around food processing areas and is suitable for cleaning air conditioner coils, refrigerators, grills, and electronic air cleaners.
  • Quickly penetrates to remove dirt, grease, and grime
  • NSF registered C1 and P1 for use in food processing areas
  • Non-corrosive and non-flammable formula
  • Foam clings to hard-to-reach areas
  • Self-rinsing on condensate-producing coils
  • Non-fuming formula with no added scent
  • Safe for overspray contact on most building, home, and roof surfaces
  • Low VOC formula
Catalog No. 50-005-02405 Supplier MSC Supplier No. 30018196
This professional-strength foaming coil cleaner penetrates quickly to remove dirt, grease, and grime that can clog coils and impede heat transfer. It is NSF registered for use in and around food processing areas and is suitable for cleaning air conditioner coils, refrigerators, grills, and electronic air cleaners.
  • Quickly penetrates to remove dirt, grease, and grime
  • NSF registered C1 and P1 for use in food processing areas
  • Non-corrosive and non-flammable formula
  • Foam clings to hard-to-reach areas
  • Self-rinsing on condensate-producing coils
  • Non-fuming formula with no added scent
  • Safe for overspray contact on most building, home, and roof surfaces
  • Low VOC formula