Learn More
Enterprise Technology Solutions Simple Green Industrial Cleaner/Degreaser, 5Gal, Clear
SDP
Catalog No. 5000830139
Encompass_Preferred
Nontoxic cleaner/degreaser features an industrial-strength formula without any added color or scents to safely clean and degrease surfaces. Versatile solution is powerful enough for intense grime while gentle enough for continued maintenance of high-traffic areas. Clean-rinsing design is excellent for surface-preparation before painting, plating or welding. Nonflammable cleaner/degreaser is perfect for use on metals, plastic, rubber, glass and more.
Catalog No. 50-008-30139 Supplier Enterprise Technology Solutions Supplier No. SMP19005
Nontoxic cleaner/degreaser features an industrial-strength formula without any added color or scents to safely clean and degrease surfaces. Versatile solution is powerful enough for intense grime while gentle enough for continued maintenance of high-traffic areas. Clean-rinsing design is excellent for surface-preparation before painting, plating or welding. Nonflammable cleaner/degreaser is perfect for use on metals, plastic, rubber, glass and more.