Learn More
Enterprise Technology Solutions Simple Green Lime Scale Remover,Deodorizer,Wintergreen Scent,1 Gal
SDP
Catalog No. 5000829129
Encompass_Preferred
Lime-Calcium-Surface Residue Remover delivers powerful cleaning and acts as a degreaser, descaler, restorer and residue remover. It is NSF registered A3 for use on food-contact surfaces, making it ideal for use in food-processing facilities and equipment. It easily removes lime scale, caustic and chlorinated cleaner residues, efflorescence, soap scum, mineral and organic deposits. This remover eliminates stains and restores clarity to stainless steel, chrome, aluminum, metals and other washable surfaces. It does not contain bleach, ammonia or phosphorus. Regular use prevents mineral deposit buildup to maintain plant asepsis.
Catalog No. 50-008-29129 Supplier Enterprise Technology Solutions Supplier No. SMP1700128
Lime-Calcium-Surface Residue Remover delivers powerful cleaning and acts as a degreaser, descaler, restorer and residue remover. It is NSF registered A3 for use on food-contact surfaces, making it ideal for use in food-processing facilities and equipment. It easily removes lime scale, caustic and chlorinated cleaner residues, efflorescence, soap scum, mineral and organic deposits. This remover eliminates stains and restores clarity to stainless steel, chrome, aluminum, metals and other washable surfaces. It does not contain bleach, ammonia or phosphorus. Regular use prevents mineral deposit buildup to maintain plant asepsis.