Learn More
Enterprise Technology Solutions Simple Green Multipurpose Cleaner, Heavy-duty, Conc, 1 Gal, Clear
SDP
Catalog No. 5000829456
Encompass_Preferred
This professional-grade, multipurpose cleaner/degreaser breaks down tough grease, grime and stains on metal, plastic, painted surfaces, sealed wood, concrete, carpet and more. Cost-effective concentrate with a 1:14 dilution ratio can be custom diluted or used in pressure washers, carpet cleaners and floor scrubbers. Noncorrosive formula is ideal for degreasing tools, equipment and vehicles. It's also perfect for use in residential and commercial construction where dirt, grime and paint overspray are common. Pro HD Heavy-duty Cleaner is safe for automotive fabrics.
Catalog No. 50-008-29456 Supplier Enterprise Technology Solutions Supplier No. SMP13421
This professional-grade, multipurpose cleaner/degreaser breaks down tough grease, grime and stains on metal, plastic, painted surfaces, sealed wood, concrete, carpet and more. Cost-effective concentrate with a 1:14 dilution ratio can be custom diluted or used in pressure washers, carpet cleaners and floor scrubbers. Noncorrosive formula is ideal for degreasing tools, equipment and vehicles. It's also perfect for use in residential and commercial construction where dirt, grime and paint overspray are common. Pro HD Heavy-duty Cleaner is safe for automotive fabrics.